missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Mouse CDCP1 Protein
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Beskrivelse
The Recombinant Mouse CDCP1 Protein is derived from NS0.
Tekniske data
Tekniske data
| Adgangsnummer | Q5U462 |
| Til brug med (applikation) | Flow Cytometry |
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gen-id (Entrez) | 64866 |
| Molekylvægt (g/mol) | 73 kDa |
| Navn | CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases |
| Mængde | 50 μg |
| Kilde | Mouse myeloma cell line,NS0-derived mouse CDCP1 protein Arg25-Ala667,with a C-terminal 6-His tag |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoksin koncentration | <1.0 EU per 1μg of the protein by the LAL method. |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion