missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD122 / IL-2RB Protein
A DNA sequence encoding the Mus musculus (Mouse) CD122 / IL-2RB, was expressed in the hosts and tags indicated.
1820.00 DKK - 19665.00 DKK
Specifications
Accession Number | NP_032394.1 |
---|---|
Gene ID (Entrez) | 16185 |
Name | CD122 / IL-2RB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16041680
|
enQuireBio™
QP6219-ye-10ug |
10 ÎĽg | N/A | ||||||
16081680
|
enQuireBio™
QP6219-ye-50ug |
50 ÎĽg | N/A | ||||||
16031680
|
enQuireBio™
QP6219-ye-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16061680
|
enQuireBio™
QP6219-ye-200ug |
200 ÎĽg | N/A | ||||||
16071680
|
enQuireBio™
QP6219-ye-500ug |
500 ÎĽg |
15875.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16051680
|
enQuireBio™
QP6219-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_032394.1 | |
CD122 / IL-2RB Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
16185 | |
Research Use Only | |
IL2RB | |
Recombinant Protein | |
Yeast | |
AVKNCSHLEC FYNSRANVSC MWSHEEALNV TTCHVHAKSN LRHWNKTCEL TLVRQASWAC NLILGSFPES QSLTSVDLLD INVVCWEEKG WRRVKTCDFH PFDNLRLVAP HSLQVLHIDT QRCNISWKVS QVSHYIEPYL EFEARRRLLG HSWEDASVLS LKQRQQWLFL EMLIPSTSYE VQVRVKAQRN NTGTWSPWSQ PLTFRTRPAD PMKE | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse CD122 / IL-2RB Protein