missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
enQuireBio™ Recombinant Mouse BAG family molecular chaperone regulator 3 Protein
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Specifications
Specifications
| Adgangsnummer | NP_038891.4 |
| Gen-id (Entrez) | 29810 |
| Navn | BAG family molecular chaperone regulator 3 Protein |
| Mængde | 50 μg |
| Regulatorisk status | Research Use Only |
| Endotoksin koncentration | Not determined. |
| Gen symbol | Bag3 |
| Protein længde | Full Length |
| Produkttype | Recombinant Protein |
| Krydsreaktivitet | Mouse |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction