missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse 14-3-3 protein zeta / delta Protein

A DNA sequence encoding the Mus musculus (Mouse) 14-3-3 protein zeta/delta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6899-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076630

  • 22245.00 DKK / 1mg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001240734.1
14-3-3 protein zeta / delta Protein
Research Use Only
Ywhaz
Recombinant Protein
E. coli
MDKNELVQKA KLAEQAERYD DMAACMKSVT EQGAELSNEE RNLLSVAYKN VVGARRSSWR VVSSIEQKTE GAEKKQQMAR EYREKIETEL RDICNDVLSL LEKFLIPNAS QPESKVFYLK MKGDYYRYLA EVAAGDDKKG IVDQSQQAYQ EAFEISKKEM QPTHPIRLGL ALNFSVFYYE ILNSPEKACS LAKTAFDEAI AELDTLSEES YKDSTLIMQL LRDNLTLWTS DTQGDEAEAG EGGEN
Greater than 90% as determined by SDS-PAGE.
22631
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers