missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque IL2 / Interleukin-2 Protein

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6217-ec-200ug

 View more versions of this product

Product Code. 16081660

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001040595.1
IL2 / Interleukin-2 Protein
Research Use Only
IL2
Recombinant Protein
E. coli
APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELKHLQCLE EELKPLEEVL NLAQSKNFHL RDTKDLISNI NVIVLELKGS ETTLMCEYAD ETATIVEFLN RWITFCQSII STLT
Greater than 82.1% as determined by SDS-PAGE.
708017
200 μg
Not determined.
Full Length of Mature Protein
Macaque
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Macaque IL2 / Interleukin-2 Protein > 200μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.