missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Macaque IL10 / Interleukin-10 Protein

Artikelnummer. 16097841
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16097841

Brand: enQuireBio™ QP9205ye1mg

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.

Specifications

Adgangsnummer NP_001038192.1
Gen-id (Entrez) 694931
Navn IL10 / Interleukin-10 Protein
Mængde 1 mg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol IL10
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Macaque
Arter Yeast
Protein tag His
Sekvens SPGQGTQSEN SCTRFPGNLP HMLRDLRDAF SRVKTFFQMK DQLDNILLKE SLLEDFKGYL GCQALSEMIQ FYLEEVMPQA ENHDPDIKEH VNSLGENLKT LRLRLRRCHR FLPCENKSKA VEQVKNAFSK LQEKGVYKAM SEFDIFINYI EAYMTMKIQN
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.