missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis Signal peptidase I Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Signal peptidase I, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7516-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_217419.1 | |
M. tuberculosis Signal peptidase I Protein | |
Research Use Only | |
lepB | |
Recombinant Protein | |
E. coli | |
RPYLIPSESM EPTLHGCSTC VGDRIMVDKL SYRFGSPQPG DVIVFRGPPS WNVGYKSIRS HNVAVRWVQN ALSFIGFVPP DENDLVKRVI AVGGQTVQCR SDTGLTVNGR PLKEPYLDPA TMMADPSIYP CLGSEFGPVT VPPGRVWVMG DNRTHSADSR AHCPLLCTDD PLPGTVPVAN VIGKARLIVW PPSRWGVVRS VNPQQGR | |
Greater than 90% as determined by SDS-PAGE. |
887157 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
M. tuberculosis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |