missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H2N2 (Korea / 426 / 1968) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/Korea/426/1968 H2N2) Influenza A H2N2 (strain A/Korea/426/1968) Hemagglutinin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7687-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
YP_308850.1 | |
Influenza A H2N2 (Korea / 426 / 1968) Hemagglutinin Protein | |
Research Use Only | |
HA | |
Recombinant Protein | |
E. coli | |
DQICIGYHAN NSTEKVDTIL ERNVTVTHAK DILEKTHNGK LCKLNGIPPL ELGDCSIAGW LLGNPECDRL LSVPEWSYIM EKENPRYSLC YPGSFNDYEE LKHLLSSVKH FEKVKILPKD RWTQHTTTGG SWACAVSGKP SFFRNMVWLT RKGSNYPVAK GSYNNTSGEQ MLIIWGVHHP NDEAEQRALY QNVGTYVSVA TSTLYKRSIP EIAARPKVNG LGRRMEFSWT LLDMWDTINF ESTGNLVAPE YGFKISKRGS SGIMKTEGTL ENCETKCQTP LGAINTTLPF HNVHPLTIGE CPKYVKSEKL VLATGLRNVP QIES | |
Greater than 90% as determined by SDS-PAGE. |
3655103 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |