missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein GLI1 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein GLI1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8750-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001153517.1 | |
Zinc finger protein GLI1 Protein | |
Research Use Only | |
GLI1 | |
Recombinant Protein | |
E. coli | |
QEPSYQSPKF LGGSQVSPSR AKAPVNTYGP GFGPNLPNHK SGSYPTPSPC HENFVVGANR ASHRAAAPPR LLPPLPTCYG PLKVGGTNPS CGHPEVGRLG GGPALYPPPE GQVCNPLDSL DLDNTQLDFV AILDEPQGLS PPPSHDQRGS SGHTPPPSGP PNMAVGNMSV LLRSLPGETE FLNSSA | |
Greater than 90% as determined by SDS-PAGE. |
2735 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |