missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 581 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 581, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6909-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057619.1 | |
Zinc finger protein 581 Protein | |
Research Use Only | |
ZNF581 | |
Recombinant Protein | |
E. coli | |
MLVLPSPCPQ PLAFSSVETM EGPPRRTCRS PEPGPSSSIG SPQASSPPRP NHYLLIDTQG VPYTVLVDEE SQREPGASGA PGQKKCYSCP VCSRVFEYMS YLQRHSITHS EVKPFECDIC GKAFKRASHL ARHHSIHLAG GGRPHGCPLC PRRFRDAGEL AQHSRVHSGE RPFQCPHCPR RFMEQNTLQK HTRWKHP | |
Greater than 90% as determined by SDS-PAGE. |
51545 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |