missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein 155 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 155, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6901-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026650

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001247415.1
Zinc finger protein 155 Protein
Research Use Only
ZNF155
Recombinant Protein
E. coli
MTTFKEAVTF KDVAVVFTEE ELGLLDPAQR KLYRDVMLEN FRNLLSVGHQ PFHQDTCHFL REEKFWMMGT ATQREGNSGG KIQTELESVP EAGAHEEWSC QQIWEQIAKD LTRSQDSIIN NSQFFENGDV PSQVEAGLPT IHTGQKPSQG GKCKQSISDV PIFDLPQQLY SEEKSYTCDE CGKSICYISA LHVHQRVHVG EKLFMCDVCG KEFSQSSHLQ THQRVHTGEK PFKCEQCGKG FSRRSALNVH RKLHTGEKPY ICEACGKAFI HDSQLKEHKR IHTGEKPFKC DICGKTFYFR SRLKSHSMVH TGEKPFRCDT CDKSFHQRSA LNRHCMVHTG EKPYRCEQCG KGFIGRLDFY KHQVVHTGEK PYNCKECGKS FRWSSCLLNH QRVHSGEKSF KCEECGKGFY TNSQLSSHQR SHSGEKPYKC EECGKGYVTK FNLDLHQRVH TGERPYNCKE CGKNFSRASS ILNHKRLHCQ KKPFKCEDCG KRLVHRTYRK DQPRDYSGEN PSKCEDCGRR YKRRLNLDIL LSLFLNDT
Greater than 90% as determined by SDS-PAGE.
7711
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers