missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 155 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 155, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6901-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001247415.1 | |
Zinc finger protein 155 Protein | |
Research Use Only | |
ZNF155 | |
Recombinant Protein | |
E. coli | |
MTTFKEAVTF KDVAVVFTEE ELGLLDPAQR KLYRDVMLEN FRNLLSVGHQ PFHQDTCHFL REEKFWMMGT ATQREGNSGG KIQTELESVP EAGAHEEWSC QQIWEQIAKD LTRSQDSIIN NSQFFENGDV PSQVEAGLPT IHTGQKPSQG GKCKQSISDV PIFDLPQQLY SEEKSYTCDE CGKSICYISA LHVHQRVHVG EKLFMCDVCG KEFSQSSHLQ THQRVHTGEK PFKCEQCGKG FSRRSALNVH RKLHTGEKPY ICEACGKAFI HDSQLKEHKR IHTGEKPFKC DICGKTFYFR SRLKSHSMVH TGEKPFRCDT CDKSFHQRSA LNRHCMVHTG EKPYRCEQCG KGFIGRLDFY KHQVVHTGEK PYNCKECGKS FRWSSCLLNH QRVHSGEKSF KCEECGKGFY TNSQLSSHQR SHSGEKPYKC EECGKGYVTK FNLDLHQRVH TGERPYNCKE CGKNFSRASS ILNHKRLHCQ KKPFKCEDCG KRLVHRTYRK DQPRDYSGEN PSKCEDCGRR YKRRLNLDIL LSLFLNDT | |
Greater than 90% as determined by SDS-PAGE. |
7711 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |