missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger matrin-type protein 1
A DNA sequence encoding the Homo sapiens (Human) Zinc finger matrin-type protein 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8082-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003692.1 | |
Zinc finger matrin-type protein 1 | |
Research Use Only | |
ZMAT5 | |
Recombinant Protein | |
E. coli | |
MGKRYFCDYC DRSFQDNLHN RKKHLNGLQH LKAKKVWYDM FRDAAAILLD EQNKRPCRKF LLTGQCDFGS NCRFSHMSER DLQELSIQVE EERRAREWLL DAPELPEGHL EDWLEKRAKR LSSAPSSRAE PIRTTVFQYP VGWPPVQELP PSLRAPPPGG WPLQPRVQWG | |
Greater than 90% as determined by SDS-PAGE. |
55954 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |