missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Wilm's Tumor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Wilm's Tumor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8720-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000369.3 | |
Wilm's Tumor 1 Protein | |
Research Use Only | |
WT1 | |
Recombinant Protein | |
E. coli | |
QGYSTVTFDG TPSYGHTPSH HAAQFPNHSF KHEDPMGQQG SLGEQQYSVP PPVYGCHTPT DSCTGSQALL LRTPYSSDNL YQMTSQLECM TWNQMNLGAT LKGVAAGSSS SVKWTEGQSN HSTGYESDNH TTPILCGAQY RIHTHGVFRG IQDVRRVPGV APTLVRSASE TSEKRPFMCA YPGCNKRYFK LSHLQMHSRK HTGEKPYQCD FKDCERRFSR SDQLKRHQRR HTGVKPFQCK TCQRKFSRSD HLKTHTRTHT GKTSEKPFSC RWPSCQKKFA RSDELVRHHN MHQRNMTKLQ | |
Greater than 90% as determined by SDS-PAGE. |
7490 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |