missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VEGFB Protein
A DNA sequence encoding the Homo sapiens (Human) VEGFB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9463-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001230662.1 | |
VEGFB Protein | |
Research Use Only | |
VEGFB | |
Recombinant Protein | |
Yeast | |
PVSQPDAPGH QRKVVSWIDV YTRATCQPRE VVVPLTVELM GTVAKQLVPS CVTVQRCGGC CPDDGLECVP TGQHQVRMQI LMIRYPSSQL GEMSLEEHSQ CECRPKKKDS AVKPDRAATP HHRPQPRSVP GWDSAPGAPS PADITHPTPA PGPSAHAAPS TTSALTPGPA AAAADAAASS VAKGGA | |
Greater than 90% as determined by SDS-PAGE. |
7423 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |