missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vasculin Protein
A DNA sequence encoding the Homo sapiens (Human) Vasculin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7795-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001120707.2 | |
Vasculin Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
65056 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
GPBP1 | |
Recombinant Protein | |
E. coli | |
MRTDKKSEFL KALKRDRVEE EHEDESRAGS EKDDDSFNLH NSNSTHQERD INRNFDENEI PQENGNASVI SQQIIRSSTF PQTDVLSSSL EAEHRLLKEM GWQEDSENDE TCAPLTEDEM REFQVISEQL QKNGLRKNGI LKNGLICDFK FGPWKNSTFK PTTENDDTET SSSDTSDDDD V |