missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UTP--glucose-1-phosphate uridylyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) UTP--glucose-1-phosphate uridylyltransferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7578-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001521.1 | |
UTP--glucose-1-phosphate uridylyltransferase Protein | |
Research Use Only | |
UGP2 | |
Recombinant Protein | |
E. coli | |
MSQDGASQFQ EVIRQELELS VKKELEKILT TASSHEFEHT KKDLDGFRKL FHRFLQEKGP SVDWGKIQRP PEDSIQPYEK IKARGLPDNI SSVLNKLVVV KLNGGLGTSM GCKGPKSLIG VRNENTFLDL TVQQIEHLNK TYNTDVPLVL MNSFNTDEDT KKILQKYNHC RVKIYTFNQS RYPRINKESL LPVAKDVSYS GENTEAWYPP GHGDIYASFY NSGLLDTFIG EGKEYIFVSN IDNLGATVDL YILNHLMNPP NGKRCEFVME VTNKTRADVK GGTLTQYEGK LRLVEIAQVP KAHVDEFKSV SKFKIFNTNN LWISLAAVKR LQEQNAIDME IIVNAKTLDG GLNVIQLETA VGAAIKSFEN SLGINVPRSR FLPVKTTSDL LLVMSNLYSL NAGSLTMSEK REFPTVPLVK LGSSFTKVQD YLRRFESIPD MLELDHLTVS GDVTFGKNVS LKGTVIIIAN HGDRIDIPPG AVLENKIVSG NLRILDH | |
Greater than 90% as determined by SDS-PAGE. |
7360 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |