Få mere at vide
enQuireBio™ Recombinant Human Uncharacterized protein C19orf48 Protein
Artikelnummer. 16080511
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16080511
Brand: enQuireBio™ QP7738ec100ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C19orf48, was expressed in the hosts and tags indicated.
Tekniske data
Adgangsnummer NP_001277078.1
Gen-id (Entrez) 84798
Navn Uncharacterized protein C19orf48 Protein
Mængde 100 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol C19orf48
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter E. coli
Protein tag GST
Sekvens MTVLEAVLEI QAITGSRLLS MVPGPARPPG SCWDPTQCTR TWLLSHTPRR RWISGLPRAS CRLGEEPPPL PYCDQAYGEE LSIRHRETWA WLSRTDTAWP GAPGVKQARI LGELLLV
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 80% as determined by SDS-PAGE.