missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UDP-N-acetylhexosamine pyrophosphorylase Protein
A DNA sequence encoding the Homo sapiens (Human) UDP-N-acetylhexosamine pyrophosphorylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7555-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001311042.1 | |
UDP-N-acetylhexosamine pyrophosphorylase Protein | |
Research Use Only | |
UAP1 | |
Recombinant Protein | |
E. coli | |
MNINDLKLTL SKAGQEHLLR FWNELEEAQQ VELYAELQAM NFEELNFFFQ KAIEGFNQSS HQKNVDARME PVPREVLGSA TRDQDQLQAW ESEGLFQISQ NKVAVLLLAG GQGTRLGVAY PKGMYDVGLP SRKTLFQIQA ERILKLQQVA EKYYGNKCII PWYIMTSGRT MESTKEFFTK HKYFGLKKEN VIFFQQGMLP AMSFDGKIIL EEKNKVSMAP DGNGGLYRAL AAQNIVEDME QRGIWSIHVY CVDNILVKVA DPRFIGFCIQ KGADCGAKVV EKTNPTEPVG VVCRVDGVYQ VVEYSEISLA TAQKRSSDGR LLFNAGNIAN HFFTVPFLRD VVNVYEPQLQ HHVAQKKIPY VDTQGQLIKP DKPNGIKMEK FVFDIFQFAK KFVVYEVLRE DEFSPLKNAD SQNGKDNPTT ARHALMSLHH CWVLNAGGHF IDENGSRLPA IPRLKDANDV PIQCEISPLI SYAGEGLESY VADKEFHAPL IIDENGVHEL VKNGI | |
Greater than 90% as determined by SDS-PAGE. |
6675 | |
200 ÎĽg | |
Not determined. | |
Full Length of Isoform AGX1. | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human UDP-N-acetylhexosamine pyrophosphorylase Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction