missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tubulin-specific chaperone C Protein
A DNA sequence encoding the Homo sapiens (Human) Tubulin-specific chaperone C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6763-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003183.1 | |
Tubulin-specific chaperone C Protein | |
Research Use Only | |
TBCC | |
Recombinant Protein | |
E. coli | |
ESVSCSAAAV RTGDMESQRD LSLVPERLQR REQERQLEVE RRKQKRQNQE VEKENSHFFV ATFVRERAAV EELLERAESV ERLEEAASRL QGLQKLINDS VFFLAAYDLR QGQEALARLQ AALAERRRGL QPKKRFAFKT RGKDAASSTK VDAAPGIPPA VESIQDSPLP KKAEGDLGPS WVCGFSNLES QVLEKRASEL HQRDVLLTEL SNCTVRLYGN PNTLRLTKAH SCKLLCGPVS TSVFLEDCSD CVLAVACQQL RIHSTKDTRI FLQVTSRAIV EDCSGIQFAP YTWSYPEIDK DFESSGLDRS KNNWNDVDDF NWLARDMASP NWSILPEEER NIQW | |
Greater than 90% as determined by SDS-PAGE. |
6903 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |