missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRMT112 Protein
A DNA sequence encoding the Homo sapiens (Human) TRMT112, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8188-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273011.1 | |
TRMT112 Protein | |
Research Use Only | |
TRMT112 | |
Recombinant Protein | |
E. coli | |
MKLLTHNLLS SHVRGVGSRG FPLRLQATEV RICPVEFNPN FVARMIPKVE WSAFLEAADN LRLIQVPKGP VEGYEENEEF LRTMHHLLLE VEVIEGTLQC PESGRMFPIS RGIPNMLLSE EETES | |
Greater than 90% as determined by SDS-PAGE. |
51504 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |