missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcription intermediary factor 1-beta Protein

A DNA sequence encoding the Homo sapiens (Human) Transcription intermediary factor 1-beta, was expressed in the hosts and tags indicated.

1185.00 DKK - 17250.00 DKK

Specifications

Accession Number NP_005753.1
Gene ID (Entrez) 10155
Name Transcription intermediary factor 1-beta Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16074971
View Documents
enQuireBio™
QP8501-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16014981
View Documents
enQuireBio™
QP8501-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16064971
View Documents
enQuireBio™
QP8501-ec-100ug
100 ÎĽg
3825.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16094971
View Documents
enQuireBio™
QP8501-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16004981
View Documents
enQuireBio™
QP8501-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16084971
View Documents
enQuireBio™
QP8501-ec-1mg
1 mg
17250.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_005753.1
Transcription intermediary factor 1-beta Protein
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
10155
Research Use Only
TRIM28
Recombinant Protein
E. coli
PGEGSAGGEK RSTAPSAAAS ASASAAASSP AGGGAEALEL LEHCGVCRER LRPEREPRLL PCLHSACSAC LGPAAPAAAN SSGDGGAAGD GTVVDCPVCK QQCFSKDIVE NYFMRDSGSK AATDAQDANQ CCTSCEDNAP ATSYCVECSE PLCETCVEAH QRVKYTKDHT VRSTGPAKSR DGERTVYCNV HKHEPLVLFC ESCDTLTCRD CQLNAHKDHQ YQFLEDAVRN QRKLLASLVK RLGDKHATLQ KSTKEVRSSI RQVSDVQKRV
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Transcription intermediary factor 1-beta Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.