missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thyroid hormone-inducible hepatic Protein
A DNA sequence encoding the Homo sapiens (Human) Thyroid hormone-inducible hepatic protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7931-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003242.1 | |
Thyroid hormone-inducible hepatic Protein | |
Research Use Only | |
Thrsp | |
Recombinant Protein | |
E. coli | |
MQVLTKRYPK NCLLTVMDRY AAEVHNMEQV VMIPSLLRDV QLSGPGGQAQ AEAPDLYTYF TMLKAICVDV DHGLLPREEW QAKVAGSEEN GTAETEEVED ESASGELDLE AQFHLHFSSL HHILMHLTEK AQEVTRKYQE MTGQVW | |
Greater than 80% as determined by SDS-PAGE. |
7069 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |