missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thyroid hormone-inducible hepatic Protein

A DNA sequence encoding the Homo sapiens (Human) Thyroid hormone-inducible hepatic protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7931-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041671

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003242.1
Thyroid hormone-inducible hepatic Protein
Research Use Only
Thrsp
Recombinant Protein
E. coli
MQVLTKRYPK NCLLTVMDRY AAEVHNMEQV VMIPSLLRDV QLSGPGGQAQ AEAPDLYTYF TMLKAICVDV DHGLLPREEW QAKVAGSEEN GTAETEEVED ESASGELDLE AQFHLHFSSL HHILMHLTEK AQEVTRKYQE MTGQVW
Greater than 80% as determined by SDS-PAGE.
7069
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers