missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Human Thiamin pyrophosphokinase 1 Protein

Artikelnummer. 16046110
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16046110

Brand: enQuireBio™ QP6827ec10ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

A DNA sequence encoding the Homo sapiens (Human) Thiamin pyrophosphokinase 1, was expressed in the hosts and tags indicated.

Tekniske data

Adgangsnummer NP_001035947.1
Gen-id (Entrez) 27010
Navn Thiamin pyrophosphokinase 1 Protein
Mængde 10 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol TPK1
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter E. coli
Protein tag GST
Sekvens MEHAFTPLEP LLSTGNLKYC LVILNQPLDN YFRHLWNKAL LRACADGGAN RLYDITEGER ESFLPEFING DFDSIRPEVR EYYATKGCEL ISTPDQDHTD FTKCLKMLQK KIEEKDLKVD VIVTLGGLAG RFDQIMASVN TLFQATHITP FPIIIIQEES LIYLLQPGKH RLHVDTGMEG DWCGLIPVGQ PCMQVTTTGL KWNLTNDVLA FGTLVSTSNT YDGSGVVTVE TDHPLLWTMA IKS
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 80% as determined by SDS-PAGE.
Vis mere Vis mindre
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.