missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGF-beta 1 / TGFB1 Protein
Recombinant Protein
Brand: enQuireBio™ QP773-20 ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000651.3 | |
TGF-beta 1 / TGFB1 Protein | |
Research Use Only | |
TGFB1 | |
Recombinant Protein | |
Human | |
ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS | |
Greater than 95% as determined by SDS-PAGE. |
7040 | |
20 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 500 pg/mL in the Inhibition of IL-4-induced HT-2 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^6 units/mg. | |
Human | |
Untagged | |
0.1 % trifluoroacetic acid (TFA) and Trehalose in a 20:1 Trehalose to protein ratio |