missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein
Beskrivelse
The Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein is derived from HEK293. The Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Adgangsnummer | P01137 |
| Til brug med (applikation) | Bioactivity |
| Gen-id (Entrez) | 7040 |
| Molekylvægt (g/mol) | 12.8 kDa (monomer) |
| Mængde | 500 μg |
| Kilde | Human embryonic kidney cell,HEK293-derived human TGF-beta 1 protein Ala279-Ser390 |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <0.01 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?