missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Recombinant Human TGF-beta 1 (Human Cell-expressed)

Artikelnummer. 18786816
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
1000 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18786816 100 μg
18789816 500 μg
18757676 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18786816 Leverandør R&D Systems™ Leverandørnr. 7754BH100/CF

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human TGF-beta 1 (Human Cell-expressed) is derived from HEK293. The Recombinant Human TGF-beta 1 (Human Cell-expressed) has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P01137
Til brug med (applikation) Bioactivity
Gen-id (Entrez) 7040
Molekylvægt (g/mol) 12.8 kDa (monomer)
Mængde 100 μg
Kilde Human embryonic kidney cell,HEK293-derived human TGF-beta 1 protein Ala279-Ser390
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatorisk status RUO
Endotoksin koncentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Gen symbol TGFB1
Specifik reaktivitet TGF-beta 1
Konjugeret Unconjugated
Arter Human
Rekombinant Recombinant
Rekonstitution Reconstitute at 100 μg/mL in sterile 4 mM HCl.
Buffer Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Renhed eller kvalitetsklasse >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.