missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ Recombinant Human TGDS GST (N-Term) Protein
Shop alle Bio Techne produkter
Klik for at se tilgængelige muligheder
Mængde:
10 μg
25 μg
Beskrivelse
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Tekniske data
Tekniske data
| Til brug med (applikation) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulering | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gen-id (Entrez) | 23483 |
| Molekylvægt (g/mol) | MolecularWeight-theroretical: 64.24 kDa |
| Mængde | 10 μg |
| Opbevaringskrav | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | dTDP-D-glucose 4,6-dehydratase, EC 4.2.1.46, growth-inhibiting protein 21, P, SDR2E1, short chain dehydrogenase/reductase family 2E, member 1, TDPGD, TDP-glucose 4,6-dehydratase |
| Gen symbol | TGDS |
| Renhed eller kvalitetsklasse | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | TGDS |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion