missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Testis-expressed sequence 12 Protein

A DNA sequence encoding the Homo sapiens (Human) Testis-expressed sequence 12 protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6773-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16085730

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_112565.1
Testis-expressed sequence 12 Protein
Research Use Only
TEX12
Recombinant Protein
E. coli
MMANHLVKPD NRNCKRPREL ESPVPDSPQL SSLGKSDSSF SEISGLFYKD EALEKDLNDV SKEINLMLST YAKLLSERAA VDASYIDEID ELFKEANAIE NFLIQKREFL RQRFTVIANT LHR
Greater than 90% as determined by SDS-PAGE.
56158
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers