missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCEANC2 Protein
A DNA sequence encoding the Homo sapiens (Human) TCEANC2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7915-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_694580.1 | |
TCEANC2 Protein | |
Research Use Only | |
TCEANC2 | |
Recombinant Protein | |
E. coli | |
MDKFVIRTPR IQNSPQKKDS GGKVYKQATI ESLKRVVVVE DIKRWKTMLE LPDQTKENLV EALQELKKKI PSREVLKSTR IGHTVNKMRK HSDSEVASLA REVYTEWKTF TEKHSNRPSI EVRSDPKTES LRKNAQKLLS EALELKMDHL LVENIERETF HLCSRLINGP YRRTVRALVF TLKHRAEIRA QVKSGSLPVG TFVQTHKK | |
Greater than 90% as determined by SDS-PAGE. |
127428 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |