missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TBCEL Protein
A DNA sequence encoding the Homo sapiens (Human) TBCEL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7704-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001123519.1 | |
TBCEL Protein | |
Research Use Only | |
TBCEL | |
Recombinant Protein | |
E. coli | |
MDQPSGRSFM QVLCEKYSPE NFPYRRGPGM GVHVPATPQG SPMKDRLNLP SVLVLNSCGI TCAGDEKEIA AFCAHVSELD LSDNKLEDWH EVSKIVSNVP QLEFLNLSSN PLNLSVLERT CAGSFSGVRK LVLNNSKASW ETVHMILQEL PDLEELFLCL NDYETVSCPS ICCHSLKLLH ITDNNLQDWT EIRKLGVMFP SLDTLVLANN HLNAIEEPDD SLARLFPNLR SISLHKSGLQ SWEDIDKLNS FPKLEEVRLL GIPLLQPYTT EERRKLVIAR LPSVSKLNGS VVTDGEREDS ERFFIRYYVD VPQEEVPFRY HELITKYGKL EPLAEVDLRP QSSAKVEVHF NDQVEEMSIR LDQTVAELKK QLKTLVQLPT SNMLLYYFDH EAPFGPEEMK YSSRALHSFG IRDGDKIYVE SKTK | |
Greater than 90% as determined by SDS-PAGE. |
219899 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |