missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Talin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Talin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8498-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006280.3 | |
Talin-1 Protein | |
Research Use Only | |
TLN1 | |
Recombinant Protein | |
Yeast | |
MLDGTVKTIM VDDSKTVTDM LMTICARIGI TNHDEYSLVR ELMEEKKEEI TGTLRKDKTL LRDEKKMEKL KQKLHTDDEL NWLDHGRTLR EQGVEEHETL LLRRKFFYSD QNVDSRDPVQ LNLLYVQARD DILNGSHPVS FDKACEFAGF QCQIQFGPHN EQKHKAGFLD LKDFLPKEYV KQKGERKIFQ AHKNCGQMSE IEAKVRYVKL ARSLKTYGVS FFLVKEKMKG KNKLVPRLLG ITKECVMRVD EKTKEVIQEW NLTNIKRWAA SPKSFTLDFG DYQDGYYSVQ TTEGEQIAQL IAGYIDII | |
Greater than 90% as determined by SDS-PAGE. |
7094 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |