missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-complex protein 1 subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) T-complex protein 1 subunit beta, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_001185771.1 |
---|---|
Gene ID (Entrez) | 10576 |
Name | T-complex protein 1 subunit beta Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987689
|
enQuireBio™
QP5779-ec-10ug |
10 ÎĽg | N/A | ||||||
15927699
|
enQuireBio™
QP5779-ec-50ug |
50 ÎĽg | N/A | ||||||
15977689
|
enQuireBio™
QP5779-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15907699
|
enQuireBio™
QP5779-ec-200ug |
200 ÎĽg | N/A | ||||||
15917699
|
enQuireBio™
QP5779-ec-500ug |
500 ÎĽg | N/A | ||||||
15997689
|
enQuireBio™
QP5779-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001185771.1 | |
T-complex protein 1 subunit beta Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10576 | |
Research Use Only | |
CCT2 | |
Recombinant Protein | |
E. coli | |
ASLSLAPVNI FKAGADEERA ETARLTSFIG AIAIGDLVKS TLGPKGMDKI LLSSGRDASL MVTNDGATIL KNIGVDNPAA KVLVDMSRVQ DDEVGDGTTS VTVLAAELLR EAESLIAKKI HPQTIIAGWR EATKAAREAL LSSAVDHGSD EVKFRQDLMN IAGTTLSSKL LTHHKDHFTK LAVEAVLRLK GSGNLEAIHI IKKLGGSLAD SYLDEGFLLD KKIGVNQPKR IENAKILIAN TGMDTDKIKI FGSRVRVDST AKVAEIEHAE KEKMKEKVER ILKHGINCFI NRQLIYNYPE QLFGAAGVMA IEHADFAGVE RLALVTGGEI ASTFDHPELV KLGSCKLIEE VMIGEDKLIH FSGVALGEAC TIVLRGATQQ ILDEAERSLH DALCVLAQTV KDSRTVYGGG CSEMLMAHAV TQLANRTPGK EAVAMESYAK ALRMLPTIIA DNAGYDSADL VAQLRAAHSE GNTTAGLDMR EGTIGDMAIL GITESFQVKR QVLLSAAEAA EVILRVDNII KAAPRKRVPD HHPC | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human T-complex protein 1 subunit beta Protein