missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human T-box transcription factor TBX15 Protein Product Code.: 16081481

enQuireBio™ Recombinant Human T-box transcription factor TBX15 Protein

Product Code. 16081481
10 μg, 10µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16081481

Brand: enQuireBio™ QP7900ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) T-box transcription factor TBX15, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001317606.1
Gene ID (Entrez) 6913
Name T-box transcription factor TBX15 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TBX15
Protein Length Full Length of Isoform 2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MSSMEEIQVE LQCADLWKRF HDIGTEMIIT KAGRRMFPAM RVKITGLDPH QQYYIAMDIV PVDNKRYRYV YHSSKWMVAG NADSPVPPRV YIHPDSLASG DTWMRQVVSF DKLKLTNNEL DDQGHIILHS MHKYQPRVHV IRKDFSSDLS PTKPVPVGDG VKTFNFPETV FTTVTAYQNQ QITRLKIDRN PFAKGFRDSG RNRTGLEAIM ETYAFWRPPV RTLTFEDFTT MQKQQGGSTG TSPTTSSTGT PSPSASSHLL SPSCSPPTFH LAPNTFNVGC RESQLCNLNL SDYPPCARSN MAALQSYPGL SDSGYNRLQS GTTSATQPSE TFMPQRTPSL ISGIPTPPSL PGNSKMEAYG GQLGSFPTSQ FQYVMQAGNA ASSSSSPHMF GGSHMQQSSY NAFSLHNPYN LYGYNFPTSP RLAASPEKLS ASQSTLLCSS PSNGAFGERQ YLPSGMEHSM HMISPSPNNQ QATNTCDGRQ YGAVPGSSSQ MSVH
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human T-box transcription factor TBX15 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.