missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Suppressor of SWI4 1 homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Suppressor of SWI4 1 homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9895-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088941

  • 4195.00 DKK / 100µg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001035754.1
Suppressor of SWI4 1 homolog Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
56342
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PPAN
Recombinant Protein
Yeast
MGQSGRSRHQ KRARAQAQLR NLEAYAANPH SFVFTRGCTG RNIRQLSLDV RRVMEPLTAS RLQVRKKNSL KDCVAVAGPL GVTHFLILSK TETNVYFKLM RLPGGPTLTF QVKKYSLVRD VVSSLRRHRM HEQQFAHPPL LVLNSFGPHG MHVKLMATMF QNLFPSINVH KVNLNTIKRC LLIDYNPDSQ ELDFRHYSIK VVPVGASRGM KKLLQEKFPN MSRLQDISEL LATGAGLSES EAEPDGDHNI TELPMS-QAV AGRGNMRAQQ SAVRLTEIGP RMTLQLIKVQ EGVGEGKVMF HSFVSKTEEE LQAILEAKEK KLRLKAQRQA QQAQNVQRKQ EQREAHRKKS LEGMKKARVG GSDEEASGIP SRTASLELGE DDDEQEDDDI EYFCQAVGEA PSEDLFPEAK QKRLAKSPGR KRKRWEMDRG RGRLCDQKFP KTKDKSQGAQ ARRGPRGASR DGGRGRGRGR PGKRVA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers