missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Suppressor of SWI4 1 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Suppressor of SWI4 1 homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9895-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035754.1 | |
Suppressor of SWI4 1 homolog Protein | |
Research Use Only | |
PPAN | |
Recombinant Protein | |
Yeast | |
MGQSGRSRHQ KRARAQAQLR NLEAYAANPH SFVFTRGCTG RNIRQLSLDV RRVMEPLTAS RLQVRKKNSL KDCVAVAGPL GVTHFLILSK TETNVYFKLM RLPGGPTLTF QVKKYSLVRD VVSSLRRHRM HEQQFAHPPL LVLNSFGPHG MHVKLMATMF QNLFPSINVH KVNLNTIKRC LLIDYNPDSQ ELDFRHYSIK VVPVGASRGM KKLLQEKFPN MSRLQDISEL LATGAGLSES EAEPDGDHNI TELPMS-QAV AGRGNMRAQQ SAVRLTEIGP RMTLQLIKVQ EGVGEGKVMF HSFVSKTEEE LQAILEAKEK KLRLKAQRQA QQAQNVQRKQ EQREAHRKKS LEGMKKARVG GSDEEASGIP SRTASLELGE DDDEQEDDDI EYFCQAVGEA PSEDLFPEAK QKRLAKSPGR KRKRWEMDRG RGRLCDQKFP KTKDKSQGAQ ARRGPRGASR DGGRGRGRGR PGKRVA | |
Greater than 90% as determined by SDS-PAGE. |
56342 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |