missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Splicing factor U2AF 35 kDa subunit Protein

A DNA sequence encoding the Homo sapiens (Human) Splicing factor U2AF 35 kDa subunit, was expressed in the hosts and tags indicated.

1185.00 DKK - 17250.00 DKK

Specifications

Accession Number NP_001020374.1
Gene ID (Entrez) 102724594
Name Splicing factor U2AF 35 kDa subunit Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16096360
View Documents
enQuireBio™
QP6861-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16036370
View Documents
enQuireBio™
QP6861-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16086360
View Documents
enQuireBio™
QP6861-ec-100ug
100 ÎĽg
3825.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16016370
View Documents
enQuireBio™
QP6861-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16026370
View Documents
enQuireBio™
QP6861-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16006370
View Documents
enQuireBio™
QP6861-ec-1mg
1 mg
17250.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001020374.1
Splicing factor U2AF 35 kDa subunit Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
102724594
Research Use Only
U2AF1
Recombinant Protein
E. coli
MAEYLASIFG TEKDKVNCSF YFKIGACRHG DRCSRLHNKP TFSQTIALLN IYRNPQNSSQ SADGLRCAVS DVEMQEHYDE FFEEVFTEME EKYGEVEEMN VCDNLGDHLV GNVYVKFRRE EDAEKAVIDL NNRWFNGQPI HAELSPVTDF REACCRQYEM GECTRGGFCN FMHLKPISRE LRRELYGRRR KKHRSRSRSR ERRSRSRDRG RGGGGGGGGG GGGRERDRRR SRDRERSGRF
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Splicing factor U2AF 35 kDa subunit Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.