missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spliceosome RNA helicase DDX39B Protein

A DNA sequence encoding the Homo sapiens (Human) Spliceosome RNA helicase DDX39B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8315-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16053961

  • 3450.00 DKK / 100µg
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_004631.1
Spliceosome RNA helicase DDX39B Protein
Research Use Only
DDX39B
Recombinant Protein
E. coli
AENDVDNELL DYEDDEVETA AGGDGAEAPA KKDVKGSYVS IHSSGFRDFL LKPELLRAIV DCGFEHPSEV QHECIPQAIL GMDVLCQAKS GMGKTAVFVL ATLQQLEPVT GQVSVLVMCH TRELAFQISK EYERFSKYMP NVKVAVFFGG LSIKKDEEVL KKNCPHIVVG TPGRILALAR NKSLNLKHIK HFILDECDKM LEQLDMRRDV QEIFRMTPHE KQVMMFSATL SKEIRPVCRK FMQDPMEIFV
Greater than 90% as determined by SDS-PAGE.
7919
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers