Få mere at vide
Novus Biologicals™ Recombinant Human SOD1/Cu-Zn SOD Protein
Artikelnummer. 18161820 Shop alle Bio Techne produkter
Skift visning
Mængde:
100 μg
Artikelnummer. Mængde
18161820 100 μg
1 options
Artikelnummer. 18161820 Leverandør Novus Biologicals™ Leverandørnr. NBP199596100UG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified and high bioactivity. Generating reliable and reproducible results.
Recombinant bioactive protein for Human SOD1/Cu-Zn SOD The Recombinant Human SOD1/Cu-Zn SOD Protein is derived from E. coli. The Recombinant Human SOD1/Cu-Zn SOD Protein has been validated for the following applications: SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY
Tekniske data
Koncentration Lyoph
Til brug med (applikation) Bioactivity, SDS-PAGE
Formulering Lyophilized.
Gen-id (Entrez) 6647
Molekylvægt (g/mol) 16.8 kDa
Mængde 100 μg
Kilde E. coli
Opbevaringskrav Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Gen symbol SOD1
Rekonstitution Reconstitute in H2O to >100 g/mL. The solution can then be diluted into other aqueous buffers.
Buffer Lyophilized.
Endotoksin niveau <0.1 ng/μg
Renhed eller kvalitetsklasse >98%, by SDS-PAGE