missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SMARCB1 Protein

A DNA sequence encoding the Homo sapiens (Human) SMARCB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7605-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16059710

  • 4195.00 DKK / 100µg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001007469.1
SMARCB1 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
6598
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SMARCB1
Recombinant Protein
Yeast
MMMALSKTFG QKPVKFQLED DGEFYMIGSE VGNYLRMFRG SLYKRYPSLW RRLATVEERK KIVASSHGKK TKPNTKDHGY TTLATSVTLL KASEVEEILD GNDEKYKAVS ISTEPPTYLR EQKAKRNSQW VPTLPNSSHH LDAVPCSTTI NRNRMGRDKK RTFPLCFDDH DPAVIHENAS QPEVLVPIRL DMEIDGQKLR DAFTWNMNEK LMTPEMFSEI LCDDLDLNPL TFVPAIASAI RQQIESYPTD SILEDQSDQR VIIKLNIHVG NISLVDQFEW DMSEKENSPE KFALKLCSEL GLGGEFVTTI AYSIRGQLSW HQKTYAFSEN PLPTVEIAIR NTGDADQWCP LLETLTDAEM EKKIRDQDRN TRRMR
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers