missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Small ubiquitin-related modifier 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Small ubiquitin-related modifier 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7696-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001002255.1 | |
Small ubiquitin-related modifier 4 Protein | |
Research Use Only | |
SUMO4 | |
Recombinant Protein | |
E. coli | |
MANEKPTEEV KTENNNHINL KVAGQDGSVV QFKIKRQTPL SKLMKAYCEP RGLSVKQIRF RFGGQPISGT DKPAQLEMED EDTIDVFQQP TGGVY | |
Greater than 80% as determined by SDS-PAGE. |
387082 | |
10 ÎĽg | |
Not determined. | |
Full Length of BC130305 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Small ubiquitin-related modifier 4 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction