Learn More
enQuireBio™ Recombinant Human Small ubiquitin-related modifier 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Small ubiquitin-related modifier 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7696-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001002255.1 | |
Small ubiquitin-related modifier 4 Protein | |
Research Use Only | |
SUMO4 | |
Recombinant Protein | |
E. coli | |
MANEKPTEEV KTENNNHINL KVAGQDGSVV QFKIKRQTPL SKLMKAYCEP RGLSVKQIRF RFGGQPISGT DKPAQLEMED EDTIDVFQQP TGGVY | |
Greater than 80% as determined by SDS-PAGE. |
387082 | |
50 μg | |
Not determined. | |
Full Length of BC130305 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |