missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Small ubiquitin-related modifier 4 Protein

A DNA sequence encoding the Homo sapiens (Human) Small ubiquitin-related modifier 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7696-ec-200ug

 View more versions of this product

Product Code. 16010261

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001002255.1
Small ubiquitin-related modifier 4 Protein
Research Use Only
SUMO4
Recombinant Protein
E. coli
MANEKPTEEV KTENNNHINL KVAGQDGSVV QFKIKRQTPL SKLMKAYCEP RGLSVKQIRF RFGGQPISGT DKPAQLEMED EDTIDVFQQP TGGVY
Greater than 80% as determined by SDS-PAGE.
387082
200 ÎĽg
Not determined.
Full Length of BC130305
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Small ubiquitin-related modifier 4 Protein > 200ÎĽg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.