Få mere at vide
enQuireBio™ Recombinant Human Signal-regulatory protein beta-1 Protein
Artikelnummer. 16085090
Mængde:
100 μg
20 μg
50 μg
Pakningsstørrelse:
100µg
20µg
50µg
Artikelnummer. 16085090
Brand: enQuireBio™ QP6685ma50ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Homo sapiens (Human) Signal-regulatory protein beta-1, was expressed in the hosts and tags indicated.
Tekniske data
Adgangsnummer NP_001077379.1
Gen-id (Entrez) 10326
Navn Signal-regulatory protein beta-1 Protein
Mængde 50 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol SIRPB1
Protein længde Partial (see sequence information for more details).
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter Mammal
Protein tag His
Sekvens EDELQVIQPE KSVSVAAGES ATLRCAMTSL IPVGPIMWFR GAGAGRELIY NQKEGHFPRV TTVSELTKRN NLDFSISISN ITPADAGTYY CVKFRKGSPD DVEFKSGAGT ELSVRAKPSA PVVSGPAVRA TPEHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP AGDSVSYSIH STARVVLTRG DVHSQVICEI AHITLQGDPL RGTANLSEAI RVPPTLEVTQ QPMRAENQAN VTCQVSNFYP RGLQLTWLEN GNVSRTETAS TLIENKDGTY NWMSWLLVNT CAHRDDVVLT CQVEHDGQQA VSKSYALEIS AHQKEHGSDI THEAALAPTA PL
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.