missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serum amyloid P component / APCS / SAP Protein
A DNA sequence encoding the Homo sapiens (Human) Serum amyloid P component / APCS / SAP, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_001630.1 |
---|---|
Gene ID (Entrez) | 325 |
Name | Serum amyloid P component / APCS / SAP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15916799
|
enQuireBio™
QP5659-ec-10ug |
10 ÎĽg | N/A | ||||||
15956799
|
enQuireBio™
QP5659-ec-50ug |
50 ÎĽg | N/A | ||||||
15906799
|
enQuireBio™
QP5659-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15936799
|
enQuireBio™
QP5659-ec-200ug |
200 ÎĽg | N/A | ||||||
15946799
|
enQuireBio™
QP5659-ec-500ug |
500 ÎĽg | N/A | ||||||
15926799
|
enQuireBio™
QP5659-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001630.1 | |
Serum amyloid P component / APCS / SAP Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
325 | |
Research Use Only | |
APCS | |
Recombinant Protein | |
E. coli | |
HTDLSGKVFV FPRESVTDHV NLITPLEKPL QNFTLCFRAY SDLSRAYSLF SYNTQGRDNE LLVYKERVGE YSLYIGRHKV TSKVIEKFPA PVHICVSWES SSGIAEFWIN GTPLVKKGLR QGYFVEAQPK IVLGQEQDSY GGKFDRSQSF VGEIGDLYMW DSVLPPENIL SAYQGTPLPA NILDWQALNY EIRGYVIIKP LVWV | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Serum amyloid P component / APCS / SAP Protein