missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Serine/arginine-rich splicing factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6679-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16085030

  • 15780.00 DKK / 1mg
Estimated Shipment: 31-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001071634.1
Serine / arginine-rich splicing factor 1 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
6426
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SRSF1
Recombinant Protein
E. coli
SGGGVIRGPA GNNDCRIYVG NLPPDIRTKD IEDVFYKYGA IRDIDLKNRR GGPPFAFVEF EDPRDAEDAV YGRDGYDYDG YRLRVEFPRS GRGTGRGGGG GGGGGAPRGR YGPPSRRSEN RVVVSGLPPS GSWQDLKDHM REAGDVCYAD VYRDGTGVVE FVRKEDMTYA VRKLDNTKFR SHEGETAYIR VKVDGPRSPS YGRSRSRSRS RSRSRSRSNS RSRSYSPRRS RGSPRYSPRH SRSRSRT
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers