missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Semenogelin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Semenogelin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6662-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002998.1 | |
Semenogelin-1 Protein | |
Research Use Only | |
SEMG1 | |
Recombinant Protein | |
E. coli | |
QKGGSKGRLP SEFSQFPHGQ KGQHYSGQKG KQQTESKGSF SIQYTYHVDA NDHDQSRKSQ QYDLNALHKT TKSQRHLGGS QQLLHNKQEG RDHDKSKGHF HRVVIHHKGG KAHRGTQNPS QDQGNSPSGK GISSQYSNTE ERLWVHGLSK EQTSVSGAQK GRKQGGSQSS YVLQTEELVA NKQQRETKNS HQNKGHYQNV VEVREEHSSK VQTSLCPAHQ DKLQHGSKDI FSTQDELLVY NKNQHQTKNL NQDQQHGRKA NKISYQSSST EERRLHYGEN GVQKDVSQSS IYSQTEEKAQ GKSQKQITIP SQEQEHSQKA NKISYQSSST EERRLHYGEN GVQKDVSQRS IYSQTEKLVA GKSQIQAPNP KQEPWHGEN | |
Greater than 90% as determined by SDS-PAGE. |
6406 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |