missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCAN domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) SCAN domain-containing protein 1, was expressed in the hosts and tags indicated.

1185.00 DKK - 17250.00 DKK

Specifications

Accession Number NP_057642.1
Gene ID (Entrez) 51282
Name SCAN domain-containing protein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16074820
View Documents
enQuireBio™
QP6651-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16014830
View Documents
enQuireBio™
QP6651-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16064820
View Documents
enQuireBio™
QP6651-ec-100ug
100 ÎĽg
3825.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16094820
View Documents
enQuireBio™
QP6651-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16004830
View Documents
enQuireBio™
QP6651-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16084820
View Documents
enQuireBio™
QP6651-ec-1mg
1 mg
17250.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_057642.1
SCAN domain-containing protein 1
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
51282
Research Use Only
SCAND1
Recombinant Protein
E. coli
MAATEPILAA TGSPAAVPPE KLEGAGSSSA PERNCVGSSL PEASPPAPEP SSPNAAVPEA IPTPRAAASA ALELPLGPAP VSVAPQAEAE ARSTPGPAGS RLGPETFRQR FRQFRYQDAA GPREAFRQLR ELSRQWLRPD IRTKEQIVEM LVQEQLLAIL PEAARARRIR RRTDVRITG
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human SCAN domain-containing protein 1

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.