missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A15 / S100A7A Protein
A DNA sequence encoding the Homo sapiens (Human) S100A15 / S100A7A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7789-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_789793.1 | |
S100A15 / S100A7A Protein | |
Research Use Only | |
S100A7A | |
Recombinant Protein | |
E. coli | |
SNTQAERSII GMIDMFHKYT GRDGKIEKPS LLTMMKENFP NFLSACDKKG IHYLATVFEK KDKNEDKKID FSEFLSLLGD IAADYHKQSH GAAPCSGGSQ | |
Greater than 90% as determined by SDS-PAGE. |
338324 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human S100A15 / S100A7A Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction